Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isocitrate Dehydrogenase 1/IDH1 Antibody (OTI3G9), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB270989A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270989A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270989A48 Supplier Novus Biologicals Supplier No. NBP270989AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Isocitrate Dehydrogenase 1/IDH1 Monoclonal specifically detects Isocitrate Dehydrogenase 1/IDH1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.

Specifications

Antigen Isocitrate Dehydrogenase 1/IDH1
Applications Western Blot, Immunohistochemistry, Immunoassay
Classification Monoclonal
Clone OTI3G9
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Knockout Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cytosolic NADP-isocitrate dehydrogenase, EC 1.1.1.42, IDCD, IDH, IDP, IDPC, isocitrate dehydrogenase [NADP] cytoplasmic, isocitrate dehydrogenase 1 (NADP+), soluble, NADP(+)-specific ICDH, NADP-dependent isocitrate dehydrogenase, cytosolic, NADP-dependent isocitrate dehydrogenase, peroxisomal, Oxalosuccinate decarboxylase, PICD
Gene Symbols IDH1
Host Species Mouse
Immunogen Synthetic peptide around the R132H mutation region of the human Isocitrate Dehydrogenase 1/IDH1 conjugated to KLH.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3417.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.