Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isocitrate Dehydrogenase 1/IDH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093046 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093046 20 μL
NB093045 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093046 Supplier Novus Biologicals Supplier No. NBP30348120UL

Rabbit Polyclonal Antibody

Isocitrate Dehydrogenase 1/IDH1 Polyclonal specifically detects Isocitrate Dehydrogenase 1/IDH1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, Knockout Validated.

Specifications

Antigen Isocitrate Dehydrogenase 1/IDH1
Applications Western Blot, Immunoprecipitation, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Cytosolic NADP-isocitrate dehydrogenase, EC 1.1.1.42, IDCD, IDH, IDP, IDPC, isocitrate dehydrogenase [NADP] cytoplasmic, isocitrate dehydrogenase 1 (NADP+), soluble, NADP(+)-specific ICDH, NADP-dependent isocitrate dehydrogenase, cytosolic, NADP-dependent isocitrate dehydrogenase, peroxisomal, Oxalosuccinate decarboxylase, PICD
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-414 of human Isocitrate Dehydrogenase 1/IDH1 (NP_005887.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3417
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.