Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl-β-D-thiogalactopyranoside, MP Biomedicals™

Catalog No. MP1IPTG0010 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
10 x 1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1IPTG0010 10 x 1 g
MP1IPTG0001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP1IPTG0010 Supplier MP Biomedicals, Inc Supplier No. 11IPTG0010
Add to Cart
Add to Cart

Isopropyl-β-D-thiogalactopyranoside Genomic Grade, >99%, White Powder

  • IPTG is a galactose analogue not recognized by β-galactosidase. It used as a Lac operon inducer in bacteria.
  • At 1mM concentration, IPTG induces the β-galactosidase gene expression in bacteria, then it allows the selection of Lac+ recombinant bacteria in X-Gal containing medium.
  • It is used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies that induce expression of the lac operon in Escherichia coli.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym IPTG, Isopropyl β-D-1-thiogalactopyranoside
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 110°C (literature)
Color White
pH 7
Quantity 10 x 1 g
Beilstein 4631
Solubility Information 10g/L in water.
Optical Rotation -28.9° to -32° (c=1, water)
Grade >99%
Physical Form Powder
storageNote1 0°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.