Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Ether (Certified ACS), Fisher Chemical™

Catalog No. p-151181 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 108-20-3, 128-37-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Boiling Range 2 °C max., Inclusive of 67 °C
Melting Point -85°C
Density .7250g/cm³
Boiling Point 68°C
Flash Point −27°C
Packaging Glass Bottle
Linear Formula [(CH3)2CH]2O
Quantity 4 L
UN Number 1159
Residue after Evaporation 0.01 % max.
Specific Gravity Inclusive Between 0.720
Stabilizer Inclusive Between 90ppm
Vapor Density 3.52g/mol
Vapor Pressure 150mmHg at 25°C
Formula Weight 102.18
Grade Certified
Physical Form Liquid
Chemical Name or Material Isopropyl Ether
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.