Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl ether, 98+%, pure, stabilized with BHT

Catalog No. p-4510359
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 108-20-3 | C6H14O | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-20-3, 128-37-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Acidity 0.005% max.
Melting Point -85.5°C
Density 0.7200g/mL
Boiling Point 68°C
Flash Point -29°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.3670 to 1.369
Linear Formula [(CH3)2CH]2O
Quantity 1 L
Beilstein 01,362
Specific Gravity 0.72
Merck Index 15,5258
Solubility Information Solubility in water: 9g/L (20°C). Other solubilities: miscible with most common organic solvents
Stabilizer 5 to 10ppm BHT
Viscosity 0.38 mPa.s (25°C)
Formula Weight 102.18
Percent Purity 98+%
Grade Pure
Physical Form Liquid
Chemical Name or Material Isopropyl ether
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Danger
EINECSNumber 203-560-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.