Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl methanesulfonate, 99%

Catalog No. p-152061
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle

CAS: 926-06-7 | C4H10O3S | 138.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926-06-7
Molecular Formula C4H10O3S
Molecular Weight (g/mol) 138.19
MDL Number MFCD00047802
InChI Key SWWHCQCMVCPLEQ-UHFFFAOYSA-N
Synonym isopropyl methanesulfonate, isopropyl mesylate, isopropylmethanesulfonate, methanesulfonic acid, 1-methylethyl ester, isopropyl methane sulphonate, 2-propyl methanesulphonate, isopropyl methane sulfonate, methanesulfonic acid, isopropyl ester, isopropyl methanesulphonate, unii-t0k2txy26b
PubChem CID 13551
IUPAC Name propan-2-yl methanesulfonate
SMILES CC(C)OS(=O)(=O)C

Specifications

Color Colorless
Density 1.1450g/mL
Boiling Point 82.0°C (6.0 mmHg)
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4180 to 1.4210
Linear Formula CH3SO3CH(CH3)2
Quantity 5 g
UN Number 1760
Beilstein 04,IV,13
Specific Gravity 1.145
Formula Weight 138.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isopropyl Methanesulfonate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-132-0
RTECSNumber PB2275000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.