Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phthalic anhydride, ACS reagent
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging:
Glass bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 129.0°C to 132.0°C |
| Color | White |
| Boiling Point | 284.0°C |
| Flash Point | 152°C |
| Ignition Residue | 0.01% max. |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Quantity | 500 g |
| Beilstein | 17,469 |
| Fieser | 01,882; 17,148 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. Causes serious eye damage. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allerg |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Do not breat |
| EINECSNumber | 201-607-5 |
| RTECSNumber | TI3150000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction