Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ISOX INACT
Click to view available options
Quantity:
10 mg
50 mg
Description
ISOX INACT is a negative control for PF CBP1 . Exhibits negligible activity at CBP and BRD4.
Active Analog also available.
Specifications
Specifications
Chemical Name or Material | 5-(Dimethyl-1,2-oxazol-4-yl)-2-[2-(4-methoxyphenyl)ethyl]--4,6-dimethyl-1-[2-(morpholin-4-yl)ethyl]-1H-1,3-benzodiazole |
Quantity | 10 mg |
Target | Bromodomain Inhibitors |
Molecular Formula | C29H36N4O3 |
Formula Weight | 488.62 |
Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction