Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ISYNA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | ISYNA1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15498420
![]() |
Novus Biologicals
NBP15498420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP154984
![]() |
Novus Biologicals
NBP154984 |
100 μL |
Each for $487.50
|
|
|||||
Description
ISYNA1 Polyclonal specifically detects ISYNA1 in Human samples. It is validated for Western Blot.Specifications
ISYNA1 | |
Polyclonal | |
Rabbit | |
Apoptosis | |
EC 5.5.1.4, hINO1, hIPS, INO1, INOS, inositol-3-phosphate synthase 1, IPSIPS 1, MI-1-P synthase, MIP synthase, Myo-inositol 1-phosphate synthase A1, Myo-inositol-1-phosphate synthase | |
ISYNA1 | |
IgG | |
61 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q9NPH2 | |
51477 | |
Synthetic peptides corresponding to ISYNA1(myo-inositol 1-phosphate synthase A1) The peptide sequence was selected from the N terminal of ISYNA1. Peptide sequence LQEQLWPHMEALRPRPSVYIPEFIAANQSARADNLIPGSRAQQLEQIRRD. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title