Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITCH/AIP4 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB188921 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188921 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB188921 Supplier Novus Biologicals Supplier No. NBP281837V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ITCH/AIP4 Polyclonal antibody specifically detects ITCH/AIP4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen ITCH/AIP4
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias AIF4, AIP4dJ468O1.1 (atrophin 1 interacting protein 4 (AIP4)), atrophin-1 interacting protein 4, Atrophin-1-interacting protein 4, dJ468O1.1, EC 6.3.2, EC 6.3.2.-, Itch, itchy (mouse homolog) E3 ubiquitin protein ligase, itchy E3 ubiquitin protein ligase homolog (mouse), itchy homolog E3 ubiquitin protein ligase, NAPP1E3 ubiquitin-protein ligase Itchy homolog, NFE2-associated polypeptide 1, ubiquitin protein ligase ITCH
Host Species Rabbit
Immunogen ITCH antibody was raised against a 15 amino acid peptide near the amino terminus of human ITCH . The immunogen is located within amino acids 40 - 90 of ITCH. Amino Acid Squence: NNTNSPKWKQPLTV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 83737
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.