Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITCH/AIP4 Rabbit anti-Human, FITC, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB089160 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB089160 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB089160 Supplier Novus Biologicals Supplier No. NBP299713F

Rabbit Polyclonal Antibody

ITCH/AIP4 Polyclonal specifically detects ITCH/AIP4 in Human samples. It is validated for ELISA.

Specifications

Antigen ITCH/AIP4
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias AIF4, AIP4dJ468O1.1 (atrophin 1 interacting protein 4 (AIP4)), atrophin-1 interacting protein 4, Atrophin-1-interacting protein 4, dJ468O1.1, EC 6.3.2, EC 6.3.2.-, Itch, itchy (mouse homolog) E3 ubiquitin protein ligase, itchy E3 ubiquitin protein ligase homolog (mouse), itchy homolog E3 ubiquitin protein ligase, NAPP1E3 ubiquitin-protein ligase Itchy homolog, NFE2-associated polypeptide 1, ubiquitin protein ligase ITCH
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ITCH/AIP4 (NP_113671.3; Arg 526-Glu 903).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83737
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.