Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITCH/AIP4 Rabbit anti-Human, Mouse, Rat, Clone: 7I7B10, Novus Biologicals™
SDP

Catalog No. NB168430 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168430 20 μg
NB168429 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168430 Supplier Novus Biologicals Supplier No. NBP31666920UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ITCH/AIP4 Monoclonal antibody specifically detects ITCH/AIP4 in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDown

Specifications

Antigen ITCH/AIP4
Applications Western Blot, KnockDown
Classification Monoclonal
Clone 7I7B10
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Knockout Validated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias AIF4, AIP4dJ468O1.1 (atrophin 1 interacting protein 4 (AIP4)), atrophin-1 interacting protein 4, Atrophin-1-interacting protein 4, dJ468O1.1, EC 6.3.2, EC 6.3.2.-, Itch, itchy (mouse homolog) E3 ubiquitin protein ligase, itchy E3 ubiquitin protein ligase homolog (mouse), itchy homolog E3 ubiquitin protein ligase, NAPP1E3 ubiquitin-protein ligase Itchy homolog, NFE2-associated polypeptide 1, ubiquitin protein ligase ITCH
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ITCH/AIP4 (Q96J02). MSDSGSQLGSMGSLTMKSQLQITVISAKLKENKKNWFGPSPYVEVTVDGQSKKTEKCNNTNSPKWKQPLTVIVTPVSKLHFRVWSHQTLKSDVLLGTAAL
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 83737
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.