Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITCH/AIP4 Rabbit anti-Human, Mouse, Rat, Clone: 7I7B10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | ITCH/AIP4 |
---|---|
Clone | 7I7B10 |
Dilution | Western Blot 1:500 - 1:2000, Knockout Validated |
Applications | Western Blot, KnockDown |
Classification | Monoclonal |
Description
ITCH/AIP4 Monoclonal antibody specifically detects ITCH/AIP4 in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDownSpecifications
ITCH/AIP4 | |
Western Blot 1:500 - 1:2000, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AIF4, AIP4dJ468O1.1 (atrophin 1 interacting protein 4 (AIP4)), atrophin-1 interacting protein 4, Atrophin-1-interacting protein 4, dJ468O1.1, EC 6.3.2, EC 6.3.2.-, Itch, itchy (mouse homolog) E3 ubiquitin protein ligase, itchy E3 ubiquitin protein ligase homolog (mouse), itchy homolog E3 ubiquitin protein ligase, NAPP1E3 ubiquitin-protein ligase Itchy homolog, NFE2-associated polypeptide 1, ubiquitin protein ligase ITCH | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ITCH/AIP4 (Q96J02). MSDSGSQLGSMGSLTMKSQLQITVISAKLKENKKNWFGPSPYVEVTVDGQSKKTEKCNNTNSPKWKQPLTVIVTPVSKLHFRVWSHQTLKSDVLLGTAAL | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
7I7B10 | |
Western Blot, KnockDown | |
Unconjugated | |
Rabbit | |
Apoptosis, Cell Biology, Immunology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
83737 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title