Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ITE
GREENER_CHOICESDP

Catalog No. 180310 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-031-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-031-0 Supplier Tocris Bioscience™ Supplier No. 1803/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Endogenous agonist for the transcription factor aryl hydrocarbon receptor

Endogenous aryl hydrocarbon receptor (AhR) agonist (Ki = 3 nM). Decreases Oct4 levels in U87 glioblastoma cells. Induces stem-like cancer cell differentiation in U87 tumor spheres and inhibits ovarian cancer cell proliferation in vitro. Suppresses tumor growth in U87 and OVCAR-3 cell xenografts in mice. Also inhibits TGF-β-induced human myofibroblast differentiation.

Chemical Identifiers

CAS 448906-42-1
Molecular Formula C14H10N2O3S

Specifications

Quantity 10 mg
Formula Weight 286.3
Purity >98%
Molecular Weight (g/mol) 286.305
Chemical Name or Material ITE
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.