Learn More
Invitrogen™ ITGA4 Monoclonal Antibody (R1-2), FITC

Description
ITGA4 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | ITGA4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | R1-2 |
| Concentration | 0.1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 4-5mg/mL BSA and 0.02% sodium azide |
| Gene | ITGA4 |
| Gene Accession No. | Q00651 |
| Gene Alias | 269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.