Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGAV Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7211671 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5561 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5561 Supplier Proteintech Group Inc Supplier No. 270961AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ITGAV belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha subunit and a beta subunit that function in cell surface adhesion and signaling. The encoded preproprotein is proteolytically processed to generate light and heavy chains that comprise the alpha V subunit. This subunit associates with beta 1, beta 3, beta 5, beta 6 and beta 8 subunits. The heterodimer consisting of alpha V and beta 3 subunits is also known as the vitronectin receptor. This integrin may regulate angiogenesis and cancer progression. Alternative splicing results in multiple transcript variants. Note that the integrin alpha 5 and integrin alpha V subunits are encoded by distinct genes.

Specifications

Antigen ITGAV
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ITGAV
Gene Accession No. P06756
Gene Alias CD51, Integrin alpha V, ITGAV, MSK8, VNRA
Gene Symbols ITGAV
Host Species Rabbit
Immunogen ITGAV Fusion Protein Ag25825
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3685
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.