Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITIH1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096289 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096289 20 μL
NB096288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096289 Supplier Novus Biologicals Supplier No. NBP30509220UL

Rabbit Polyclonal Antibody

ITIH1 Polyclonal specifically detects ITIH1 in Human samples. It is validated for Western Blot.

Specifications

Antigen ITIH1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias H1P, IATIH, IGHEP1, inter-alpha (globulin) inhibitor H1, inter-alpha (globulin) inhibitor, H1 polypeptide, Inter-alpha-inhibitor heavy chain 1, Inter-alpha-trypsin inhibitor complex component III, inter-alpha-trypsin inhibitor heavy chain H1, ITI heavy chain H1, ITIH, ITI-HC1, MGC126415, Serum-derived hyaluronan-associated protein, SHAP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 530-670 of human ITIH1 (NP_002206.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3697
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.