Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITIH3 Antibody (049), HRP, Novus Biologicals™
SDP

Catalog No. NB072331 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072331 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072331 Supplier Novus Biologicals Supplier No. NBP290329H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ITIH3 Monoclonal specifically detects ITIH3 in Human samples. It is validated for ELISA.

Specifications

Antigen ITIH3
Applications ELISA
Classification Monoclonal
Clone 49
Conjugate HRP
Dilution ELISA
Formulation PBS
Gene Alias H3 polypeptide, inter-alpha (globulin) inhibitor H3, inter-alpha (globulin) inhibitor, H3 polypeptide, inter-alpha-inhibitor heavy chain 3, inter-alpha-trypsin inhibitor heavy chain H3, ITI heavy chain H3, ITI-HC3, serum-derived hyaluronan-associated protein, SHAP
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ITIH3 (NP_002208.3; Met1-Asp651).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3699
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.