Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITK Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB267337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB267337 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB267337 Supplier Novus Biologicals Supplier No. NBP232240H

Rabbit Polyclonal Antibody

ITK Polyclonal antibody specifically detects ITK in Human samples. It is validated for Western Blot

Specifications

Antigen ITK
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias EC 2.7.10, EC 2.7.10.2, EMTMGC126257, homolog of mouse T-cell itk/tsk, IL2-inducible T-cell kinase, Kinase EMT, LYKMGC126258, PSCTK2, T-cell-specific kinase, tyrosine-protein kinase ITK/TSK, Tyrosine-protein kinase Lyk
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human IL2-Inducible T-cell Kinase using the numbering given in entry NP_005537.3 (GeneID 3702).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3702
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.