Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITPA Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB169608 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB169608 100 μg
NB169609 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB169608 Supplier Novus Biologicals Supplier No. NBP317266100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ITPA Polyclonal antibody specifically detects ITPA in Human samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen ITPA
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias C20orf37, dJ794I6.3, EC 3.6.1, EC 3.6.1.19, HLC14-06-P, Inosine triphosphatase, inosine triphosphatase (nucleoside triphosphate pyrophosphatase), inosine triphosphatase-A, inosine triphosphate pyrophosphatase, inosine triphosphate pyrophosphohydrolase, ITPase, My049 protein, nucleoside triphosphate diphosphatase, Putative oncogene protein hlc14-06-p
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: GNAKKLEEVVQILGDKFPCTLVAQKIDLPEYQGEPDEISIQKCQEAVRQVQGPVLVEDTCLCFNALGGLPGPYIKWFLEKLKPEGLHQL
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3704
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.