Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITPK1 Antibody (OTI1F5), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289131 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289131 Supplier Novus Biologicals Supplier No. NBP271052APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ITPK1 Monoclonal antibody specifically detects ITPK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen ITPK1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1F5
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias EC 2.7.1.134, EC 2.7.1.159, inositol 13,4-triphosphate 5/6 kinase, inositol 13,4-trisphosphate 5/6 kinase, Inositol 13,4-trisphosphate 5/6-kinase, inositol-tetrakisphosphate 1-kinase, Inositol-triphosphate 5/6-kinase, Ins(13,4)P(3) 5/6-kinase, ITRPK1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 309-391 of human ITPK1(NP_001136065) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3705
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.