Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IVD Rabbit anti-Human, DyLight 550, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB065128 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB065128 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB065128 Supplier Novus Biologicals Supplier No. NBP300257R

Rabbit Polyclonal Antibody

IVD Polyclonal specifically detects IVD in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IVD
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias ACAD2, EC 1.3.99, EC 1.3.99.10, FLJ12715, FLJ34849, isovaleryl-CoA dehydrogenase, mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant IVD (AAH17202.1; His30-His423).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3712
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.