Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IVD Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB170346 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170346 50 μg
NB170366 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170346 Supplier Novus Biologicals Supplier No. NBP31879850UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IVD Polyclonal antibody specifically detects IVD in Human samples. It is validated for ELISA

Specifications

Antigen IVD
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:5000 - 1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias ACAD2, EC 1.3.99, EC 1.3.99.10, FLJ12715, FLJ34849, isovaleryl-CoA dehydrogenase, mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified recombinant Human IVD (His30-His423, Accession: AAH17202.1)
Purification Method Antigen and protein A Affinity-purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3712
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.