Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ivosidenib
GREENER_CHOICE

Catalog No. p-7249493
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent inhibitor of mutant isocitrate dehydrogenase (mIDH1)

Ivosidenib is a potent inhibitor of mutant isocitrate dehydrogenase enzyme 1 (mIDH1), with activity against a variety of mutant IDH1 enzymes (IC50 range = 5-19 nM). Reduces intracellular 2-HG (Cat. No. 6122) by 96% at 0.5 μM. Increases sensitivity of melanoma to Temozolomide (Cat. No. 2706) in vivo and inhibits wild-type IDH1 in low magnesium and low nutrient levels. Ivosidenib also inhibits growth of tumors in cell and animal models of pancreatic cancer and inhibits invasion and migration of chondrosarcoma cells in vitro.

Specifications

Chemical Name or Material (2S)-N-{(1S)-1-(2-Chlorophenyl)-2-[(3,3-difluorocyclobutyl)amino]-2-oxoethyl}-1(4-cyanopyridin-2-yl)-N-(5-fluoropyridin-3-yl)-5-oxopyrrolidine-2-carboxamide
CAS 1448347-49-6
Quantity 10 mg
Target Isocitrate Dehydrogenase 1/IDH1
Molecular Formula C28H22ClF3N6O3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.