Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Western Devices Benchtop Bundle

Catalog No. iw3000s
Encompass_Preferred
Click to view available options
Type:
Western Devices Bundle

Invitrogen Western Devices Benchtop bundle contains all the necessary devices, consumables and reagents to perform high quality westerns

The Western Devices Benchtop Bundle contains all the necessary devices, consumables, and reagents to perform high quality westerns.

Contents of the Western Devices Benchtop Bundle:
• Mini Gel Tank (Cat. No. A25977)
• Bolt 4-12% Bis-Tris Plus gels, 10-well (Cat. No. NW04120BOX)
• Bolt MES Running Buffer, 20X (Cat. No. B0002)
• Bolt LDS Sample Buffer, 4X (Cat. No. B0007)
• Bolt Sample Reducing Agent, 10X (Cat. No. B0009)
• SeeBlue Plus2 Pre-stained Protein Standard (Cat. No. LC5925)
• iBlot 2 Gel Transfer Device (Cat. No. IB21001)
• iBlot 2 Transfer Stacks, nitrocellulose, regular size (Cat. No. IB23001)
• iBind Western Device (Cat. No. SLF1000)
• iBind Cards (Cat. No. SLF1010)
• iBind Solution Kit (Cat. No. SLF1020)
• SuperSignal West Pico Plus Chemiluminescent Substrate (Cat. No. 34577)

Mini Gel Tank
The Mini Gel Tank is compatible with Novex gels, NuPAGE gels, and Bolt Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt Bis-Tris Plus gels to be run in approximately 35 minutes.

Bolt Bis-Tris Plus gels
Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of a wide range of protein sizes under denaturing conditions. Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt gels to obtain optimal results for your day-to-day protein separation needs.

iBlot 2 Gel Transfer Device
The iBlot 2 Gel Transfer Device performs western blotting transfer simply, efficiently, and reliably, within seven minutes without the need for liquid buffers. The iBlot 2 system is compatible with both polyvinylidene difluoride (PVDF) and nitrocellulose membranes, and has comparable performance to traditional wet transfer methods in a fraction of the time. The iBlot 2 Gel Transfer Device is an integral part of the iBlot 2 Dry Blotting system, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane.

iBind Western Device
The iBind Western Device is an automated western-processing device that performs every step in the western protocol from blocking to washes to antibody incubations via sequential lateral flow (SLF). SLF allows the timely release and flow of solutions and antibodies to the membrane without need of an external power source. Each solution is released from the iBind device wells to an iBind Card via SLF; the solutions are then wicked towards the stack region of the card. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction. The iBind western detection procedure minimizes hands-on time and is highly reproducible and compatible with all western protocols. Set up time is fast: in less than 3 hours you are ready for the detection step.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Mini Gel Tank
• Bolt 4-12% Bis-Tris Plus gels, 10-well
• Bolt MES Running Buffer (20X)
• Bolt LDS Sample Buffer (4X)
• Bolt Sample Reducing Agent (10X)
• SeeBlue Plus2 Pre-stained Protein Standard
• iBlot 2 Gel Transfer Device
• iBlot 2 Transfer Stacks, nitrocellulose, regular size
• iBind Western Device
• iBind Cards
• iBind Solution Kit
• SuperSignal West Pico Plus Chemiluminescent Substrate

Detection Method Fluorescence, Chemiluminescence, Chromogenic
For Use With (Application) Western Blot
Gel Compatibility Bis-Tris Gel, E-PAGE™ Gel, Midi, Mini, Tricine Gel, Tris-Acetate Gel, Tris-Glycine Gel
Membrane Compatibility Nitrocellulose, PVDF
Mode of Transfer Dry
Product Line iBlot 2
Type Western Devices Bundle
Quantity 1 each
Shipping Condition Approved for shipment at Room Temperature and wet ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.