Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IZUMO1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB185187 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185187 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185187 Supplier Novus Biologicals Supplier No. NBP282097JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IZUMO1 Polyclonal antibody specifically detects IZUMO1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen IZUMO1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias FLJ61440, IZUMO, izumo sperm-egg fusion 1, izumo sperm-egg fusion protein 1, MGC34799, OBF, Oocyte binding/fusion factor, Sperm-specific protein izumo
Host Species Rabbit
Immunogen IZUMO1 antibody was raised against an 18 amino acid peptide near the center of human IZUMO1. The immunogen is located within amino acids 170 - 220 of IZUMO1. Amino Acid Squence: ELNWHQASEGLTDYSFY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284359
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.