Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ J 113863
GREENER_CHOICE

Catalog No. 25951
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-951 1 mg
25-951-0 10 mg
25-955-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 25-951 Supplier Tocris Bioscience™ Supplier No. 2595/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent CCR1 chemokine receptor antagonist

Potent chemokine receptor 1 (CCR1) antagonist (IC50 values are 0.9 and 5.8 nM for human and mouse CCR1 receptors respectively). Also displays high selectivity for human but not mouse CCR3 receptors (IC50 values are 0.58 and 460 nM respectively). Improves paw inflammation, joint damage and dramatically reduces cell infiltration into joints in collagen-induced arthritis in mice.

Isomer also available.

Chemical Identifiers

CAS 353791-85-2
Molecular Formula C30H37Cl2IN2O2

Specifications

Quantity 1 mg
Formula Weight 655.44
Purity >98%
Molecular Weight (g/mol) 655.442
Chemical Name or Material J 113863
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.