Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

J Chain Antibody (Mc19-9), Novus Biologicals™
SDP

Catalog No. NB10064825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064825 0.1 mg
NB017652 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064825 Supplier Novus Biologicals Supplier No. NB10064825

Mouse Monoclonal Antibody

J Chain Monoclonal specifically detects J Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen J Chain
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone Mc19-9
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 20 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 20 ug/ml, Immunohistochemistry-Frozen 1:200
Host Species Mouse
Immunogen Human J Chain
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64825 reacts with human J chain as tested by ELISA using purified human Ig fragments as the solid phase target antigen. The antibody does not detect J chain in intact IgM in ELISA assays due to steric hindrance. In Western blotting techniques NB100-64825 detects J chain in IgM only in reducing conditions.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.