Learn More
Stoelting™ Jacobson Mosquito Forceps
SDP
Stoelting™ Jacobson Mosquito Forceps are used in many surgical procedures usually to control bleeding or the flow of liquids in tubing.
$475.00
Specifications
For Use With (Application) Neuroscience
Length (English) 5 in.
View More Specs
Products 2
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-001-326
SDP
View Documents
Stoelting™
5211006
Straight, Extra-delicate
Each for $475.00
Only null left
Add to Cart
10-001-327
SDP
View Documents
Stoelting™
5211007
Curved, Extra-delicate
Each for $475.00
Only null left
Add to Cart
Description
Description
  • Scissor-like ring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
  • Delicate hemostats have thin jaws with fine serrations.
Specifications
Specifications
Neuroscience
5 in.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.