Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Jade Bottle Top Dispenser
SDPSCI_ED

Catalog No. S139575 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Volume (Metric):
0.25 to 2.5 mL
0.5 to 5 mL
1 to 10 mL
10 to 100 mL
2.5 to 30 mL
5 to 60 mL
Increments:
0.05 mL
0.1 mL
0.2 mL
0.5 mL
1 mL
2 mL
Weight (English):
1.5 lb.
1.75 lb.
2 lb.
2.25 lb.
4 lb.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Increments Weight (English)
S139575 1 to 10 mL 0.2 mL 1.75 lb.
S139573 0.25 to 2.5 mL 0.05 mL 1.5 lb.
S139574 0.5 to 5 mL 0.1 mL 1.5 lb.
S139576 2.5 to 30 mL 0.5 mL 2 lb.
S139577 5 to 60 mL 1 mL 2.25 lb.
S139578 10 to 100 mL 2 mL 4 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. S139575 Supplier United Scientific™ Supplier No. BTJD10
Add to Cart
Add to Cart
This item is not returnable. View return policy

This bottle top dispenser features a recirculation valve.

Dispense a wide range of reagents with precision and accuracy using the Jade Bottle Top Dispenser.

  • Bottle top dispensers are available in a variety of a volume ranges
  • Five adapters are included with each unit for use with a wide range of bottle sizes
  • Adapters included are: 28 mm, 32 mm, 38 mm, 40 mm and 45 mm
  • Features a recirculation valve that redirects liquid back into the bottle without the loss of reagent
  • Operates with a smooth plunger movement for bubble-free dispensing and effortless performance, even under demanding laboratory conditions
  • Fully Autoclavable at 121°C, 15 psi for 10 to 15 minutes

Specifications

Accuracy 0.005
Product Type Analog Bottletop Dispenser with Adapters
Autoclavable Autoclavable
Certifications/Compliance ISO 8655
Color White
Channels 1
Material PTFE
Volume (Metric) 1 to 10 mL
Display Analog
For Use With (Application) Dispensing Liquids
For Use With (Equipment) Chemicals
Includes Dispenser and Adapters
Increments 0.2 mL
Sterility Non-sterile
Warranty 1 Year
Weight (English) 1.75 lb.
No. per Pack 1/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.