Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jagged 1 Antibody (188331), HRP, Novus Biologicals™
SDP

Catalog No. NB350042 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350042 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350042 Supplier Novus Biologicals Supplier No. FAB1277H

Mouse Monoclonal Antibody

Jagged 1 Monoclonal antibody specifically detects Jagged 1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Jagged 1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 188331
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, jagged 1, Jagged1, JAGL1, protein jagged-1
Host Species Mouse
Immunogen NS0-derived recombinant human Jagged 1, Ser32-Asp296, Accession # P78504
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Growth and Development, Neuronal Cell Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 182
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.