Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jagged 1 Antibody (188331), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB314647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314647 Supplier Novus Biologicals Supplier No. FAB1277PEATT594

Mouse Monoclonal Antibody

Jagged 1 Monoclonal antibody specifically detects Jagged 1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry

Specifications

Antigen Jagged 1
Applications Flow Cytometry
Classification Monoclonal
Clone 188331
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, jagged 1, Jagged1, JAGL1, protein jagged-1
Host Species Mouse
Immunogen NS0-derived recombinant human Jagged 1, Ser32-Asp296, Accession # P78504
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Growth and Development, Neuronal Cell Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 182
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.