Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jagged 1 Antibody (OTI3A10), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP270115C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270115C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270115C Supplier Novus Biologicals Supplier No. NBP270115C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Jagged 1 Monoclonal specifically detects Jagged 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Jagged 1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3A10
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, jagged 1, Jagged1, JAGL1, protein jagged-1
Gene Symbols JAG1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 523-759 of human Jagged 1 (NP_000205) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer, Growth and Development, Neuronal Cell Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 182.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.