Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jagged 2 Antibody (746513), FITC, Novus Biologicals™
SDP

Catalog No. NB369085 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369085 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369085 Supplier Novus Biologicals Supplier No. FAB4748F
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Jagged 2 Monoclonal specifically detects Jagged 2 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Jagged 2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 746513
Conjugate FITC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias HJ2, jagged 2, Jagged2, protein jagged-2, SER2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Jagged 2, Met27-Leu1084, Accession # Q9QYE5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3714
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.