Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAGN1 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB190883 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB190883 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB190883 Supplier Novus Biologicals Supplier No. NBP281910DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JAGN1 Polyclonal antibody specifically detects JAGN1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen JAGN1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias FLJ14602, GL009, jagunal homolog 1 (Drosophila), protein jagunal homolog 1
Host Species Rabbit
Immunogen JAGN1 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human JAGN1. The immunogen is located within the last 50 amino acids of JAGN1. Amino Acid Squence: YSKKLLDSWFTSTQEKKHK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 84522
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.