Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jak1 Antibody (413104), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB303611 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303611 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303611 Supplier Novus Biologicals Supplier No. FAB4260U

Rat Monoclonal Antibody

Jak1 Monoclonal antibody specifically detects Jak1 in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Jak1
Applications Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 413104
Conjugate Alexa Fluor 350
Dilution Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.10, JAK-1, JAK1Atyrosine-protein kinase JAK1, JAK1B, Janus kinase 1EC 2.7.10.2, JTK3
Host Species Rat
Immunogen E. coli-derived recombinant human Jak1, Pro32-Phe286, Accession # P23458
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3716
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.