Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jak2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343397 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343397 Supplier Novus Biologicals Supplier No. NBP241310MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Jak2 Polyclonal antibody specifically detects Jak2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Jak2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.10, EC 2.7.10.2, JAK-2, Janus kinase 2Janus kinase 2 (a protein tyrosine kinase), JTK10, tyrosine-protein kinase JAK2
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human JAK2. The immunogen is located within the last 50 amino acids of JAK2. Amino Acid Squence: WNNNVNQRPSFRDLALR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3717
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.