Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jak2 Antibody (OTI7D12), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB271056A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271056A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271056A48 Supplier Novus Biologicals Supplier No. NBP271056AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Jak2 Monoclonal specifically detects Jak2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen Jak2
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI7D12
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.10, EC 2.7.10.2, JAK-2, Janus kinase 2Janus kinase 2 (a protein tyrosine kinase), JTK10, tyrosine-protein kinase JAK2
Gene Symbols JAK2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 297-596 of human Jak2 (NP_004963) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3717.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.