Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jak3 Antibody (452524), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB303470 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303470 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303470 Supplier Novus Biologicals Supplier No. FAB46991T

Mouse Monoclonal Antibody

Jak3 Monoclonal antibody specifically detects Jak3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Jak3
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 452524
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.10, JAK3_HUMAN, JAK-3EC 2.7.10.2, JAKLtyrosine-protein kinase JAK3, Janus kinase 3Leukocyte janus kinase, LJAK, L-JAKJanus kinase 3 (a protein tyrosine kinase, leukocyte)
Host Species Mouse
Immunogen E. coli-derived recombinant human Jak3, Gly46-Thr209, Accession # P52333
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3718
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.