Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jak3 Antibody (5H2), Novus Biologicals™
SDP

Catalog No. NBP237737 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237737 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237737 Supplier Novus Biologicals Supplier No. NBP237737
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Jak3 Monoclonal specifically detects Jak3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Jak3
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 5H2
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000
Formulation Ascites with 0.03% Sodium Azide
Gene Alias EC 2.7.10, JAK3_HUMAN, JAK-3EC 2.7.10.2, JAKLtyrosine-protein kinase JAK3, Janus kinase 3Leukocyte janus kinase, LJAK, L-JAKJanus kinase 3 (a protein tyrosine kinase, leukocyte)
Gene Symbols JAK3
Host Species Mouse
Immunogen Purified recombinant fragment of human JAK3 expressed in E. Coli.
Molecular Weight of Antigen 125 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3718
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.