Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jak3 Antibody (5H2), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$487.50
Specifications
Antigen | Jak3 |
---|---|
Clone | 5H2 |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
Jak3 Monoclonal specifically detects Jak3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Jak3 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
Monoclonal | |
Ascites | |
RUO | |
Human, Mouse | |
EC 2.7.10, JAK3_HUMAN, JAK-3EC 2.7.10.2, JAKLtyrosine-protein kinase JAK3, Janus kinase 3Leukocyte janus kinase, LJAK, L-JAKJanus kinase 3 (a protein tyrosine kinase, leukocyte) | |
JAK3 | |
IgG1 | |
Unpurified |
5H2 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cell Biology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
Ascites with 0.03% Sodium Azide | |
3718 | |
Purified recombinant fragment of human JAK3 expressed in E. Coli. | |
Primary | |
125 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title