Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAKMIP2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB343766 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343766 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343766 Supplier Novus Biologicals Supplier No. NBP241202MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JAKMIP2 Polyclonal antibody specifically detects JAKMIP2 in Human, Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen JAKMIP2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias CTCL tumor antigen HD-CL-04, JAMIP2KIAA0555Jak and microtubule interacting protein 2, janus kinase and microtubule interacting protein 2, janus kinase and microtubule-interacting protein 2, NECC1, neuroendocrine long coiled-coil 1, Neuroendocrine long coiled-coil protein 1
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the center of human JAKMIP2. The immunogen is located within amino acids 250 - 300 of JAKMIP2. Amino Acid Squence: RAGDGSEHCSSPDLR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9832
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.