Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAKMIP2 Antibody, Novus Biologicals™
SDP

Catalog No. NB035263 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB035263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB035263 Supplier Novus Biologicals Supplier No. NBP287641
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JAKMIP2 Polyclonal specifically detects JAKMIP2 in Human samples. It is validated for Western Blot.

Specifications

Antigen JAKMIP2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias CTCL tumor antigen HD-CL-04, JAMIP2KIAA0555Jak and microtubule interacting protein 2, janus kinase and microtubule interacting protein 2, janus kinase and microtubule-interacting protein 2, NECC1, neuroendocrine long coiled-coil 1, Neuroendocrine long coiled-coil protein 1
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of human JAKMIP2
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9832
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.