Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAM-A Antibody (RM0116-1M21), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP122494UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122494UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122494UV Supplier Novus Biologicals Supplier No. NBP122494UV
Add to Cart
Add to Cart

Rat Monoclonal Antibody

JAM-A Monoclonal specifically detects JAM-A in Mouse samples. It is validated for Western Blot.

Specifications

Antigen JAM-A
Applications Western Blot
Classification Monoclonal
Clone RM0116-1M21
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1
Gene Symbols F11R
Host Species Rat
Immunogen Mouse recombinant protein of the extracellular domain of JAM-A
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 50848
Test Specificity This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of extracellular domain of JAM-A.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.