Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAM-A Rat anti-Mouse, Clone: RM0116-1M21, Novus Biologicals™
SDP

Catalog No. NB018441 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018441 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB018441 Supplier Novus Biologicals Supplier No. NBP1224940.025MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

JAM-A Monoclonal specifically detects JAM-A in Mouse samples. It is validated for Western Blot.

Specifications

Antigen JAM-A
Applications Western Blot
Classification Monoclonal
Clone RM0116-1M21
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation A 0.2 um filtered solution in PBS
Gene Alias CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1
Host Species Rat
Immunogen Mouse recombinant protein of the extracellular domain of JAM-A
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50848
Reconstitution Reconstitute with 0.2 mL sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.