Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAM-B/VE-JAM Antibody (988905) [DyLight 405], Novus Biologicals™
SDP

Catalog No. NB401173 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401173 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401173 Supplier Novus Biologicals Supplier No. FAB10744E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

JAM-B/VE-JAM Monoclonal antibody specifically detects JAM-B/VE-JAM in Human samples. It is validated for Flow Cytometry, Neutralization

Specifications

Antigen JAM-B/VE-JAM
Applications Flow Cytometry, Neutralization
Classification Monoclonal
Clone 988905
Conjugate DyLight 405
Dilution Flow Cytometry, Neutralization
Formulation 50mM Sodium Borate
Gene Alias C21orf43, CD322 antigen, JAMB, JAM-BJAM-IT/VE-JAM, junctional adhesion molecule 2JAM-2, junctional adhesion molecule B, PRO245, Vascular endothelial junction-associated molecule, VE-JAMVEJAMCD322
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human JAM-B/VE-JAM, Phe29-Asn236, Accession # P57087
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58494
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.