Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAM-B/VE-JAM Antibody (MM0425-4L28), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP211745UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211745UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211745UV Supplier Novus Biologicals Supplier No. NBP211745UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

JAM-B/VE-JAM Monoclonal specifically detects JAM-B/VE-JAM in Human samples. It is validated for Western Blot.

Specifications

Antigen JAM-B/VE-JAM
Applications Western Blot
Classification Monoclonal
Clone MM0425-4L28
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C21orf43, CD322 antigen, JAMB, JAM-BJAM-IT/VE-JAM, junctional adhesion molecule 2JAM-2, junctional adhesion molecule B, PRO245, Vascular endothelial junction-associated molecule, VE-JAMVEJAMCD322
Gene Symbols JAM2
Host Species Mouse
Immunogen Human recombinant JAM-B/VE-JAM
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 58494
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.