Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAM-C Antibody (208212) [Alexa Fluor™ 532], Novus Biologicals™
SDP

Catalog No. NB400688 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400688 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400688 Supplier Novus Biologicals Supplier No. FAB11891X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

JAM-C Monoclonal antibody specifically detects JAM-C in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen JAM-C
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 208212
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias JAM-2, JAMC, JAM-CFLJ14529, junctional adhesion molecule 3JAM-3, junctional adhesion molecule C
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human JAM-C, Val32-Asn241(Ala149Pro), Accession # Q9BX67
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 83700
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.