Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAMP Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP236446UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236446UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP236446UV Supplier Novus Biologicals Supplier No. NBP236446UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JAMP Polyclonal specifically detects JAMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen JAMP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9P055
Gene Alias C14orf100, chromosome 14 open reading frame 100, HSPC213, HSPC327, JAMPCDA06, JNK1/MAPK8-associated membrane protein, JNK1-associated membrane protein, JNK-associated membrane protein, Jun N-terminal kinase 1-associated membrane protein, Medulloblastoma antigen MU-MB-50.4
Gene Symbols JKAMP
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human JAMP protein (between amino acids 20-150) [UniProt Q9P055]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Unfolded Protein Response (UPR)
Primary or Secondary Primary
Gene ID (Entrez) 51528
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.