Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Japanese Encephalitis virus NS1 Antibody (GT1448), Novus Biologicals™
SDP

Catalog No. NB177255 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177255 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB177255 Supplier Novus Biologicals Supplier No. NBP313568
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Japanese Encephalitis virus NS1 Monoclonal antibody specifically detects Japanese Encephalitis virus NS1 in Virus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)

Specifications

Antigen Japanese Encephalitis virus NS1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GT1448
Conjugate Unconjugated
Dilution Western Blot 0.05-0.5 ug/ml, Flow Cytometry 1:20-1:2000, Immunohistochemistry 1:10-1:500, Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular)
Formulation PBS
Gene Alias Japanese Encephalitis
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of NS1 (JEV). (Japanese Encephalitis Virus strain Jaoars982) The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489713
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.